Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dibromofluorene, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
16433-88-8
C13H8Br2
324.015
MFCD00019048
AVXFJPFSWLMKSG-UHFFFAOYSA-N
2,7-dibromofluorene,9h-fluorene, 2,7-dibromo,fluorene, 2,7-dibromo,fluorene,7-dibromo,2, 7-dibromofluorene,2,7-di bromo fluorene,9h-fluorene,7-dibromo,acmc-1c8sn,ksc491i1j
140073
2,7-dibromo-9H-fluorene
C1C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br
16433-88-8
C13H8Br2
324.015
MFCD00019048
AVXFJPFSWLMKSG-UHFFFAOYSA-N
2,7-dibromofluorene,9h-fluorene, 2,7-dibromo,fluorene, 2,7-dibromo,fluorene,7-dibromo,2, 7-dibromofluorene,2,7-di bromo fluorene,9h-fluorene,7-dibromo,acmc-1c8sn,ksc491i1j
140073
2,7-dibromo-9H-fluorene
C1C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
Melting Point 164.0°C to 167.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 324.01
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 2, 7-Dibromofluorene
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning