Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',7'-Dichlorofluorescein, ACS reagent, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
76-54-0
C20H10Cl2O5
401.195
VFNKZQNIXUFLBC-UHFFFAOYSA-N
2',7'-Dichloro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one
64944
CHEBI:51596
2',7'-dichloro-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)Cl)O)O)Cl
76-54-0
C20H10Cl2O5
401.195
VFNKZQNIXUFLBC-UHFFFAOYSA-N
2',7'-Dichloro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one
64944
CHEBI:51596
2',7'-dichloro-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)Cl)O)O)Cl
Formula Weight 401.2
Grade ACS Reagent
Packaging Glass bottle
Density 0.7900g/mL
Quantity 1g
Chemical Name or Material 2', 7'-Dichlorofluorescein