Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2′,7′-Dichlorofluorescein, ACS, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
500 g
Packaging:
Amber Glass Bottle
76-54-0
76-54-0
CAS Max % 1
Grade ACS
Packaging Amber Glass Bottle
Quantity 5 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.