Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dihydroxynaphthalene, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
582-17-2
C10H8O2
160.17
MFCD00004085
DFQICHCWIIJABH-UHFFFAOYSA-N
2,7-dihydroxynaphthalene,2,7-naphthalenediol,naphthalenediol-2,7,unii-0to8e448ud,naphthalenediol-2,7 french,2,7-dihydroxy naphthalene,2,7-naphthalene diol,naphthalene-2-7-diol,2,7-dihdroxynaphthalene
11397
naphthalene-2,7-diol
C1=CC(=CC2=C1C=CC(=C2)O)O
582-17-2
C10H8O2
160.17
MFCD00004085
DFQICHCWIIJABH-UHFFFAOYSA-N
2,7-dihydroxynaphthalene,2,7-naphthalenediol,naphthalenediol-2,7,unii-0to8e448ud,naphthalenediol-2,7 french,2,7-dihydroxy naphthalene,2,7-naphthalene diol,naphthalene-2-7-diol,2,7-dihdroxynaphthalene
11397
naphthalene-2,7-diol
C1=CC(=CC2=C1C=CC(=C2)O)O
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Melting Point 184°C to 189°C
Color Gray
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C10H6(OH)2
Beilstein 06, 985
Formula Weight 160.17
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 2, 7-Dihydroxynaphthalene
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning