Alfa Aesar™ 2,7-Dimethoxynaphthalene, 98%

Manufacturer: Alfa Aesar™ H2797114

Quantity 25g

 View more versions of this product

Catalog No. AAH2797114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3469-26-9
    Molecular Formula C12H12O2
    Formula Weight 188.23
    MDL Number MFCD00004064
    Chemical Name or Material 2,7-Dimethoxynaphthalene

    Specifications

    Formula Weight 188.23
    Molecular Formula C12H12O2
    MDL Number MFCD00004064
    CAS 3469-26-9
    Melting Point 137° to 140°C
    Quantity 25g
    Chemical Name or Material 2,7-Dimethoxynaphthalene
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit