Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,7-Dimethyl-9-fluorenone-4-carboxylic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
500536-41-4
C16H12O3
252.269
MFCD04039780
ATPRNYZONXAEOW-UHFFFAOYSA-N
2,7-dimethyl-9-oxo-9h-fluorene-4-carboxylic acid,2,7-dimethyl-9-fluorenone-4-carboxylic acid
246697
2,7-dimethyl-9-oxofluorene-4-carboxylic acid
CC1=CC2=C(C=C1)C3=C(C=C(C=C3C2=O)C)C(=O)O
500536-41-4
C16H12O3
252.269
MFCD04039780
ATPRNYZONXAEOW-UHFFFAOYSA-N
2,7-dimethyl-9-oxo-9h-fluorene-4-carboxylic acid,2,7-dimethyl-9-fluorenone-4-carboxylic acid
246697
2,7-dimethyl-9-oxofluorene-4-carboxylic acid
CC1=CC2=C(C=C1)C3=C(C=C(C=C3C2=O)C)C(=O)O
Quantity 1g
Formula Weight 252.27
Percent Purity 97%
Chemical Name or Material 2,7-Dimethyl-9-fluorenone-4-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning