Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,8-Bis(trifluoromethyl)-4-hydroxyquinoline, 97%

Click to view available options
Quantity:
1g
5g
35853-41-9
C11H5F6NO
281.157
MFCD00075091
JIWHKBAFGFPZKM-UHFFFAOYSA-N
2,8-bis trifluoromethyl-4-hydroxyquinoline,2,8-bis trifluoromethyl quinolin-4-ol,2,8-bis trifluoromethyl-4-quinolinol,2,8-bis trifluoromethyl-1h-quinolin-4-one,2,8-bis-trifluoromethyl-quinolin-4-ol,cc-pmlsc-dma-p104,4-quinolinol, 2,8-bis trifluoromethyl,2,8-bis trifluoromethyl-4-hydroxy quinoleine,2,8-di trifluoromethyl quinolin-4-ol,pubchem5934
736140
2,8-bis(trifluoromethyl)-1H-quinolin-4-one
C1=CC2=C(C(=C1)C(F)(F)F)NC(=CC2=O)C(F)(F)F
35853-41-9
C11H5F6NO
281.157
MFCD00075091
JIWHKBAFGFPZKM-UHFFFAOYSA-N
2,8-bis trifluoromethyl-4-hydroxyquinoline,2,8-bis trifluoromethyl quinolin-4-ol,2,8-bis trifluoromethyl-4-quinolinol,2,8-bis trifluoromethyl-1h-quinolin-4-one,2,8-bis-trifluoromethyl-quinolin-4-ol,cc-pmlsc-dma-p104,4-quinolinol, 2,8-bis trifluoromethyl,2,8-bis trifluoromethyl-4-hydroxy quinoleine,2,8-di trifluoromethyl quinolin-4-ol,pubchem5934
736140
2,8-bis(trifluoromethyl)-1H-quinolin-4-one
C1=CC2=C(C(=C1)C(F)(F)F)NC(=CC2=O)C(F)(F)F
Melting Point 129°C to 135°C
Quantity 1g
Formula Weight 281.16
Percent Purity 97%
Chemical Name or Material 2,8-Bis(trifluoromethyl)-4-hydroxyquinoline

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning