Alfa Aesar™ 2,9-Dimethyl-1,10-phenanthroline hemihydrate, 98+%

Manufacturer: Alfa Aesar™ A1139806

Quantity 5g

 View more versions of this product

Catalog No. AAA1139806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 34302-69-7
    Molecular Formula C14H12N2·0.5H2O
    Formula Weight 217.27 (208.26 Anhydrous)
    MDL Number MFCD00149306
    Beilstein 153179
    Merck Index 14,6449
    Synonym Neocuproine
    Chemical Name or Material 2,9-Dimethyl-1,10-phenanthroline hemihydrate

    Specifications

    Formula Weight 217.27 (208.26 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Merck Index 14,6449
    Health Hazard 1 Warning
    TSCA TSCA
    Molecular Formula C14H12N2·0.5H2O
    MDL Number MFCD00149306
    Beilstein 153179
    Quantity 5g
    Synonym Neocuproine
    Melting Point 160° to 164°C
    CAS 34302-69-7
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 2,9-Dimethyl-1,10-phenanthroline hemihydrate
    Assay ≥98%

    Description

    Applications
    2,9-Dimethyl-1,10-phenanthroline hemihydrate, is used as a reagent for the spectrophotometric determination of copper. It is used as an intermediate in medicine and chemical research.

    Solubility
    It is soluble in methanol: 0.1 g/mL.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit