Alfa Aesar™ 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonic acid disodium salt

For the colorimetric determination of copper

Manufacturer: Alfa Aesar™ 4288902

Quantity 0.5g

 View more versions of this product

Catalog No. AA4288902

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 52698-84-7
    Molecular Formula C12H4N2(CH3)2(C6H4SO3Na)2
    Formula Weight 564.57
    MDL Number MFCD00011679
    Synonym Bathocuproine disulfonic acid, disodium salt
    Chemical Name or Material 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonic acid disodium salt

    Specifications

    Formula Weight 564.57
    Molecular Formula C12H4N2(CH3)2(C6H4SO3Na)2
    Quantity 0.5g
    Physical Form Powder
    Melting Point 300°C
    MDL Number MFCD00011679
    CAS 52698-84-7
    Synonym Bathocuproine disulfonic acid, disodium salt
    Solubility Information Soluble in water
    TSCA TSCA
    Chemical Name or Material 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonic acid disodium salt

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit