Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Acetamido-4-(4-bromophenyl)thiazole, 97%

Click to view available options
Quantity:
250mg
1g
53173-91-4
C11H9BrN2OS
297.17
CALREGGMKYUGIP-UHFFFAOYSA-N
n-4-4-bromophenyl-1,3-thiazol-2-yl acetamide,2-acetamido-4-4-bromophenyl thiazole,2-n-acetamino-4-4-bromophenyl thiazole,n-4-4-bromophenyl thiazol-2-yl acetamide
790490
N-[4-(4-bromophenyl)-1,3-thiazol-2-yl]acetamide
CC(=O)NC1=NC(=CS1)C2=CC=C(C=C2)Br
53173-91-4
C11H9BrN2OS
297.17
CALREGGMKYUGIP-UHFFFAOYSA-N
n-4-4-bromophenyl-1,3-thiazol-2-yl acetamide,2-acetamido-4-4-bromophenyl thiazole,2-n-acetamino-4-4-bromophenyl thiazole,n-4-4-bromophenyl thiazol-2-yl acetamide
790490
N-[4-(4-bromophenyl)-1,3-thiazol-2-yl]acetamide
CC(=O)NC1=NC(=CS1)C2=CC=C(C=C2)Br
Quantity 1g
Melting Point 233°C to 234°C
Color White to Yellow
Formula Weight 297.17
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2-Acetamido-4-(4-bromophenyl)thiazole