Alfa Aesar™ 2-Acetamidothiazole, 98%

Manufacturer: Alfa Aesar™ B2387322

Quantity 100g

 View more versions of this product

Catalog No. AAB2387322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2719-23-5
    Molecular Formula C5H6N2OS
    Formula Weight 142.18
    MDL Number MFCD00016892
    Chemical Name or Material 2-Acetamidothiazole

    Specifications

    Formula Weight 142.18
    Melting Point 202° to 204°C
    Quantity 100g
    TSCA TSCA
    Molecular Formula C5H6N2OS
    MDL Number MFCD00016892
    CAS 2719-23-5
    RTECS Number AC8800000
    Chemical Name or Material 2-Acetamidothiazole
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit