Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-3-ethylpyrazine, 98%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
32974-92-8
C8H10N2O
150.181
MFCD00038028
PPJSYGVFDJEMRP-UHFFFAOYSA-N
2-acetyl-3-ethylpyrazine, 1-3-ethylpyrazin-2-yl ethanone, ethanone, 1-3-ethylpyrazinyl, 2-acetyl-3-ethyl-1,4-diazine, 1-3-ethylpyrazinyl ethan-1-one, unii-aq03n14xtl, 1-3-ethylpyrazinyl ethanone, 1-3-ethyl-2-pyrazinyl ethanone, 3-ethyl-2-acetylpyrazine, 2-acetyl-3-ethyl pyrazine
61918
1-(3-ethylpyrazin-2-yl)ethanone
CCC1=NC=CN=C1C(=O)C
32974-92-8
C8H10N2O
150.181
MFCD00038028
PPJSYGVFDJEMRP-UHFFFAOYSA-N
2-acetyl-3-ethylpyrazine, 1-3-ethylpyrazin-2-yl ethanone, ethanone, 1-3-ethylpyrazinyl, 2-acetyl-3-ethyl-1,4-diazine, 1-3-ethylpyrazinyl ethan-1-one, unii-aq03n14xtl, 1-3-ethylpyrazinyl ethanone, 1-3-ethyl-2-pyrazinyl ethanone, 3-ethyl-2-acetylpyrazine, 2-acetyl-3-ethyl pyrazine
61918
1-(3-ethylpyrazin-2-yl)ethanone
CCC1=NC=CN=C1C(=O)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Density 1.075
Boiling Point 54°C to 56°C (1mmHg)
Flash Point 87°C (188°F)
Beilstein 742901
Refractive Index 1.515
Quantity 5g
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 2-Acetyl-3-ethylpyrazine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 251-316-2

TSCA : Yes

Recommended Storage : Ambient temperatures