Alfa Aesar™ 2-Acetyl-5-chlorothiophene, 99%

Manufacturer: Alfa Aesar™ A1185030

Quantity 250g

 View more versions of this product

Catalog No. AAA1185030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6310-09-4
    Molecular Formula C6H5ClOS
    Formula Weight 160.62
    MDL Number MFCD00005444
    UN Number UN2811
    Beilstein 113936
    Chemical Name or Material 2-Acetyl-5-chlorothiophene

    Specifications

    Formula Weight 160.62
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00005444
    CAS 6310-09-4
    Beilstein 113936
    Health Hazard 1 Danger
    Flash Point 108°C (226°F)
    UN Number UN2811
    Melting Point 44° to 48°C
    Health Hazard 2 GHS H Statement
    H301-H312-H332
    Toxic if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number OB1745000
    Molecular Formula C6H5ClOS
    Boiling Point 117° to 118°C (17mmHg)
    Quantity 250g
    Chemical Name or Material 2-Acetyl-5-chlorothiophene
    Assay 99%

    GHS H Statement
    H301-H312-H332
    Toxic if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit