Alfa Aesar™ 2-Acetyl-5-methoxybenzo[b]furan, 97+%

Manufacturer: Alfa Aesar™ H5236403

Quantity 1g

 View more versions of this product

Catalog No. AAH5236403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21587-39-3
    Molecular Formula C11H10O3
    Formula Weight 190.2
    MDL Number MFCD01716424
    Synonym 1-(5-Methoxy-2-benzo[b]furanyl)ethanone
    Chemical Name or Material 2-Acetyl-5-methoxybenzo[b]furan

    Specifications

    Formula Weight 190.2
    MDL Number MFCD01716424
    Synonym 1-(5-Methoxy-2-benzo[b]furanyl)ethanone
    Quantity 1g
    Molecular Formula C11H10O3
    CAS 21587-39-3
    Chemical Name or Material 2-Acetyl-5-methoxybenzo[b]furan
    Assay ≥97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit