Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-5-methylfuran, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
1193-79-9
C7H8O2
124.139
MFCD00003243
KEFJLCGVTHRGAH-UHFFFAOYSA-N
2-acetyl-5-methylfuran,5-methyl-2-acetylfuran,1-5-methylfuran-2-yl ethanone,1-5-methyl-2-furyl ethanone,5-methyl-2-furylmethylketone,ethanone, 1-5-methyl-2-furanyl,1-5-methyl-2-furanyl ethanone,2-methyl-5-acetylfuran,1-5-methyl-2-furyl ethan-1-one,methyl 5-methyl-2-furyl ketone
14514
CHEBI:562752
1-(5-methylfuran-2-yl)ethanone
CC1=CC=C(O1)C(=O)C
1193-79-9
C7H8O2
124.139
MFCD00003243
KEFJLCGVTHRGAH-UHFFFAOYSA-N
2-acetyl-5-methylfuran,5-methyl-2-acetylfuran,1-5-methylfuran-2-yl ethanone,1-5-methyl-2-furyl ethanone,5-methyl-2-furylmethylketone,ethanone, 1-5-methyl-2-furanyl,1-5-methyl-2-furanyl ethanone,2-methyl-5-acetylfuran,1-5-methyl-2-furyl ethan-1-one,methyl 5-methyl-2-furyl ketone
14514
CHEBI:562752
1-(5-methylfuran-2-yl)ethanone
CC1=CC=C(O1)C(=O)C
Quantity 25g
Color Undesignated
Infrared Spectrum Authentic
Refractive Index 1.5120 to 1.5140 (20°C, 589nm)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 124.14
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Acetyl-5-methylfuran, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 214-779-1

RTECSNumber : LT8528000

TSCA : TSCA