Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-5-methylthiophene 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
13679-74-8
C7H8OS
140.2
MFCD00014529
YOSDTJYMDAEEAZ-UHFFFAOYSA-N
2-acetyl-5-methylthiophene,1-5-methylthiophen-2-yl ethanone,ketone, methyl 5-methyl-2-thienyl,1-5-methyl-2-thienyl ethan-1-one,2-methyl-5-acetylthiophene,ethanone, 1-5-methyl-2-thienyl,methylthienylcetone,1-5-methyl-2-thienyl ethanone,5-methyl-2-acetylthiophene,methyl 5-methyl-2-thienyl ketone
83655
1-(5-methylthiophen-2-yl)ethanone
CC1=CC=C(S1)C(=O)C
13679-74-8
C7H8OS
140.2
MFCD00014529
YOSDTJYMDAEEAZ-UHFFFAOYSA-N
2-acetyl-5-methylthiophene,1-5-methylthiophen-2-yl ethanone,ketone, methyl 5-methyl-2-thienyl,1-5-methyl-2-thienyl ethan-1-one,2-methyl-5-acetylthiophene,ethanone, 1-5-methyl-2-thienyl,methylthienylcetone,1-5-methyl-2-thienyl ethanone,5-methyl-2-acetylthiophene,methyl 5-methyl-2-thienyl ketone
83655
1-(5-methylthiophen-2-yl)ethanone
CC1=CC=C(S1)C(=O)C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
Color White to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 140.2
Physical Form Low Melting Crystalline Mass, Powder or Crystals
Percent Purity ≥97.5%
Chemical Name or Material 2-Acetyl-5-methylthiophene
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning