Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-5-norbornene, (Tech.) Mixture of Endo and Exo, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
5083-03-4
C14H12FNO
229.25
MFCD00167570
RXYSKAOGUKNBOB-UHFFFAOYSA-N
n-4-fluorophenyl-4-methylbenzamide,benzamide, n-4-fluorophenyl-4-methyl,4'-fluoro-4-methylbenzanilide,n~1~-4-fluorophenyl-4-methylbenzamide,n-4-fluorophenyl 4-methylphenyl carboxamide
531826
N-(4-fluorophenyl)-4-methylbenzamide
CC1=CC=C(C=C1)C(=O)NC2=CC=C(C=C2)F
5083-03-4
C14H12FNO
229.25
MFCD00167570
RXYSKAOGUKNBOB-UHFFFAOYSA-N
n-4-fluorophenyl-4-methylbenzamide,benzamide, n-4-fluorophenyl-4-methyl,4'-fluoro-4-methylbenzanilide,n~1~-4-fluorophenyl-4-methylbenzamide,n-4-fluorophenyl 4-methylphenyl carboxamide
531826
N-(4-fluorophenyl)-4-methylbenzamide
CC1=CC=C(C=C1)C(=O)NC2=CC=C(C=C2)F
Color Brown-Yellow to Yellow
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 136.19
Physical Form Liquid
Grade Technical
Chemical Name or Material 2-Acetyl-5-norbornene