Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-6-methoxypyridine 97.0+%, TCI America™
SDP

Catalog No. A28025G
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 21190-93-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD08276836
InChI Key XPJALZRCJQKJKB-UHFFFAOYSA-N
PubChem CID 13642086
IUPAC Name 1-(6-methoxypyridin-2-yl)ethan-1-one
SMILES COC1=CC=CC(=N1)C(C)=O

Specifications

Melting Point 41°C
Boiling Point 85°C
Quantity 5 g
Formula Weight 151.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetyl-6-methoxypyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.