Alfa Aesar™ 2-Acetylanthracene, 97%

Manufacturer: Alfa Aesar™ 4475106

Quantity 5g

 View more versions of this product

Catalog No. AA4475106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10210-32-9
    Molecular Formula C16H12O
    Formula Weight 220.27
    MDL Number MFCD00156727
    Chemical Name or Material 2-Acetylanthracene

    Specifications

    Formula Weight 220.27
    Molecular Formula C16H12O
    MDL Number MFCD00156727
    Quantity 5g
    Physical Form Powder
    CAS 10210-32-9
    Melting Point 188° to 192°C
    Chemical Name or Material 2-Acetylanthracene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit