Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Acetylbenzoic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
2-Acetylbenzoic acid is used as a component of hair dyes. It is used as an intermediate for pharmaceutical, cosmetics, agrochemical and food industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Acetylbenzoic acid is used as a component of hair dyes. It is used as an intermediate for pharmaceutical, cosmetics, agrochemical and food industries.
Solubility
Soluble in Dichloromethane and Ethyl Acetate. Slightly soluble in water.
Notes
Store at -20°C. Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
2-Acetylbenzoic acid is used as a component of hair dyes. It is used as an intermediate for pharmaceutical, cosmetics, agrochemical and food industries.
Solubility
Soluble in Dichloromethane and Ethyl Acetate. Slightly soluble in water.
Notes
Store at -20°C. Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications
Melting Point | 115°C to 118°C |
Odor | Odorless |
Quantity | 10 g |
Beilstein | 1210557 |
Solubility Information | Soluble in Dichloromethane and Ethyl Acetate. Slightly soluble in water. |
Formula Weight | 164.16 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Acetylbenzoic acid |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 209-413-2 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction