Alfa Aesar™ 2-Acetylbenzoic acid, 98+%

Manufacturer: Alfa Aesar™ A1021609

Quantity 10g

 View more versions of this product

Catalog No. AAA1021609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 577-56-0
    Molecular Formula C9H8O3
    Formula Weight 164.16
    MDL Number MFCD00002475
    Beilstein 1210557
    Synonym Acetophenone-2'-carboxylic acid; 3-Hydroxy-3-methylphthalide
    Chemical Name or Material 2-Acetylbenzoic acid

    Specifications

    Formula Weight 164.16
    Molecular Formula C9H8O3
    Beilstein 1210557
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    MDL Number MFCD00002475
    CAS 577-56-0
    Melting Point 115° to 118°C
    Quantity 10g
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Synonym Acetophenone-2'-carboxylic acid; 3-Hydroxy-3-methylphthalide
    Chemical Name or Material 2-Acetylbenzoic acid
    Assay ≥98%

    Description

    Applications
    2-Acetylbenzoic acid is used as a component of hair dyes. It is used as an intermediate for pharmaceutical, cosmetics, agrochemical and food industries.

    Solubility
    Soluble in Dichloromethane and Ethyl Acetate. Slightly soluble in water.

    Notes
    Store at -20°C. Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit