Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetylbenzoic Acid 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
577-56-0
C9H8O3
164.16
MFCD00002475
QDAWXRKTSATEOP-UHFFFAOYSA-N
o-acetylbenzoic acid,benzoic acid, 2-acetyl,2-acetylbenzoicacid,acetophenone-2-carboxylic acid,2-acetyl-benzoic acid,benzoic acid, acetyl,3-hydroxy-3-methylphthalide,2'-acetophenonecarboxylic acid,acetophenone-2'-carboxylic acid,methyl phenyl ketone-o-carboxylic acid
68474
2-acetylbenzoic acid
CC(=O)C1=CC=CC=C1C(=O)O
577-56-0
C9H8O3
164.16
MFCD00002475
QDAWXRKTSATEOP-UHFFFAOYSA-N
o-acetylbenzoic acid,benzoic acid, 2-acetyl,2-acetylbenzoicacid,acetophenone-2-carboxylic acid,2-acetyl-benzoic acid,benzoic acid, acetyl,3-hydroxy-3-methylphthalide,2'-acetophenonecarboxylic acid,acetophenone-2'-carboxylic acid,methyl phenyl ketone-o-carboxylic acid
68474
2-acetylbenzoic acid
CC(=O)C1=CC=CC=C1C(=O)O
Quantity 25g
Color White-Yellow
Melting Point 118°C
Formula Weight 164.16
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 2-Acetylbenzoic Acid