Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetylcyclohexanone, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
874-23-7
C8H12O2
140.18
MFCD00001633
OEKATORRSPXJHE-UHFFFAOYSA-N
2-acetylcyclohexanone,cyclohexanone, 2-acetyl,2-acetyl-1-cyclohexanone,.alpha.-acetylcyclohexanone,2-acetyl cyclohexanone,2-acetyl-cyclohexanone,cyclohexanone,2-acetyl,1-acetyl-2-oxocyclohexane,alpha-acetylcyclohexanone,acetylcyclohexanone
13400
2-acetylcyclohexan-1-one
CC(=O)C1CCCCC1=O
874-23-7
C8H12O2
140.18
MFCD00001633
OEKATORRSPXJHE-UHFFFAOYSA-N
2-acetylcyclohexanone,cyclohexanone, 2-acetyl,2-acetyl-1-cyclohexanone,.alpha.-acetylcyclohexanone,2-acetyl cyclohexanone,2-acetyl-cyclohexanone,cyclohexanone,2-acetyl,1-acetyl-2-oxocyclohexane,alpha-acetylcyclohexanone,acetylcyclohexanone
13400
2-acetylcyclohexan-1-one
CC(=O)C1CCCCC1=O
Density 1.0630g/mL
Boiling Point 224°C to 226°C
Flash Point 79°C
Specific Gravity 1.063
Refractive Index 1.5090 to 1.511
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3COC6H9(=O)
Beilstein 07, 559
Formula Weight 140.18
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Acetylcyclohexanone, 97%
Show More Show Less

EINECSNumber : 212-858-5

TSCA : TSCA