Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetylcyclopentanone, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1670-46-8
C7H10O2
126.155
MFCD00001413
OSWDNIFICGLKEE-UHFFFAOYSA-N
2-acetylcyclopentanone,cyclopentanone, 2-acetyl,o-acetylcyclopentanone,alpha-acetylcyclopentanone,acetylcyclopentanone,2-acetyl cyclopentanone,2-acetyl-cyclopentanon,2-acetyl-cyclopentanone,acmc-209dvh,cyclopentanone,2-acetyl
98471
2-acetylcyclopentan-1-one
CC(=O)C1CCCC1=O
1670-46-8
C7H10O2
126.155
MFCD00001413
OSWDNIFICGLKEE-UHFFFAOYSA-N
2-acetylcyclopentanone,cyclopentanone, 2-acetyl,o-acetylcyclopentanone,alpha-acetylcyclopentanone,acetylcyclopentanone,2-acetyl cyclopentanone,2-acetyl-cyclopentanon,2-acetyl-cyclopentanone,acmc-209dvh,cyclopentanone,2-acetyl
98471
2-acetylcyclopentan-1-one
CC(=O)C1CCCC1=O
Color Yellow to Brown
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4890 to 1.4910 (20°C, 589nm)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3COC5H7(=O)
Beilstein 07, 558
Formula Weight 126.15
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2-Acetylcyclopentanone
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning