Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Acetylfluorene, 98%

Manufacturer:  Alfa Aesar™ B2284022

Quantity 100g

 View more versions of this product

Catalog No. AAB2284022


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

781-73-7
208.26
1871711
2-Acetylfluorene
C15H12O
MFCD00001124
2-Fluorenyl methyl ketone

Specifications

781-73-7
208.26
1871711
127° to 132°C
217°C (422°F)
2-Acetylfluorene
C15H12O
MFCD00001124
2-Fluorenyl methyl ketone
100g
Odorless
98%

Safety and Handling

SDS & Safety Documents

RTECSNumber : OB3800000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit