Alfa Aesar™ 2-Acetylfluorene, 98%

Manufacturer: Alfa Aesar™ B2284014

Quantity 25g

 View more versions of this product

Catalog No. AAB2284014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 781-73-7
    Molecular Formula C15H12O
    Formula Weight 208.26
    MDL Number MFCD00001124
    Beilstein 1871711
    Synonym 2-Fluorenyl methyl ketone
    Chemical Name or Material 2-Acetylfluorene

    Specifications

    Formula Weight 208.26
    Melting Point 127° to 132°C
    Flash Point 217°C (422°F)
    RTECS Number OB3800000
    Molecular Formula C15H12O
    MDL Number MFCD00001124
    CAS 781-73-7
    Beilstein 1871711
    Synonym 2-Fluorenyl methyl ketone
    Quantity 25g
    Odor Odorless
    Chemical Name or Material 2-Acetylfluorene
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit