Alfa Aesar™ 2-Acetylquinoxaline, 97%

Manufacturer: Alfa Aesar™ H26680MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH26680MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25594-62-1
    Molecular Formula C10H8N2O
    Formula Weight 172.19
    MDL Number MFCD07808559
    Chemical Name or Material 2-Acetylquinoxaline

    Specifications

    Formula Weight 172.19
    Molecular Formula C10H8N2O
    MDL Number MFCD07808559
    CAS 25594-62-1
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Melting Point 76° to 79°C
    Quantity 250mg
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 2-Acetylquinoxaline
    Assay 97%

    Description

    Applications
    2-Acetylquinoxaline is used as dyes, pharmaceuticals, and antibiotics such as olaquindox, carbadox, echinomycin, levomycin and actinoleutin. It is also used to make amiloride.

    Solubility
    Soluble in water.

    Notes
    Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit