Alfa Aesar™ 2-Allyl-N-Boc-D-glycine dicyclohexylamine salt, 95%

Manufacturer: Alfa Aesar™ H52127MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52127MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 170899-08-8
    Molecular Formula C10H17NO4·C12H23N
    Formula Weight 396.57
    MDL Number MFCD01321012
    Chemical Name or Material 2-Allyl-N-Boc-D-glycine dicyclohexylamine salt

    Specifications

    Formula Weight 396.57
    Molecular Formula C10H17NO4·C12H23N
    MDL Number MFCD01321012
    CAS 170899-08-8
    Quantity 250mg
    Chemical Name or Material 2-Allyl-N-Boc-D-glycine dicyclohexylamine salt
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit