Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Allyl-N-Boc-L-glycine dicyclohexylamine salt, 95%

Manufacturer:  Alfa Aesar™ H5206203

Quantity 1g

 View more versions of this product

Catalog No. AAH5206203


  Check Availability
Add to cart

Specifications

Description

Applications
2-Allyl-N-Boc-L-glycine dicyclohexylamine salt is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

143979-15-1
396.57
Boc-allyl-Gly-OH (dicyclohexylammonium) salt
C10H17NO4·C12H23N
MFCD01321013
2-Allyl-N-Boc-L-glycine dicyclohexylamine salt

Specifications

143979-15-1
396.57
Boc-allyl-Gly-OH (dicyclohexylammonium) salt
2-Allyl-N-Boc-L-glycine dicyclohexylamine salt
C10H17NO4·C12H23N
MFCD01321013
1g
95%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit