Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Allyl-N-Fmoc-DL-glycine, 98%

Click to view available options
Quantity:
5g
25g
221884-63-5
C20H19NO4
337.375
MFCD02259489
YVBLQCANYSFEBN-UHFFFAOYSA-N
fmoc-alpha-allyl-dl-glycine,fmoc-dl-allylglycine,2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-enoic acid,s-n-fmoc-allyl-glycine,fmocdlallylglycine,fmoc-allyl-dl-gly-oh,fmoc-alpha-allyl-dl-gly-oh,2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-enoic acid,2-allyl-n-fmoc-dl-glycine,fmoc-2-amino-4-pentenoic acid;fmoc-allylglycine
4052587
2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-enoic acid
C=CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
221884-63-5
C20H19NO4
337.375
MFCD02259489
YVBLQCANYSFEBN-UHFFFAOYSA-N
fmoc-alpha-allyl-dl-glycine,fmoc-dl-allylglycine,2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-enoic acid,s-n-fmoc-allyl-glycine,fmocdlallylglycine,fmoc-allyl-dl-gly-oh,fmoc-alpha-allyl-dl-gly-oh,2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-enoic acid,2-allyl-n-fmoc-dl-glycine,fmoc-2-amino-4-pentenoic acid;fmoc-allylglycine
4052587
2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-enoic acid
C=CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Formula Weight 337.37
Physical Form Solid
Percent Purity 98%
Quantity 5g
Chemical Name or Material 2-Allyl-N-Fmoc-DL-glycine

storageNote1 : 2° to 8°C