Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Allyloxybenzaldehyde 90%, ACROS Organics™

Click to view available options
Quantity:
25mL
Packaging:
Glass bottle
28752-82-1
C10H10O2
162.188
MFCD00014130
BXCJDECTRRMSCV-UHFFFAOYSA-N
2-allyloxy benzaldehyde,2-allyloxybenzaldehyde,o-allyloxy benzaldehyde,o-allyloxybenzaldehyde,benzaldehyde, 2-2-propenyloxy,benzaldehyde, o-allyloxy,2-prop-2-en-1-yloxy benzaldehyde,2-prop-2-enyloxybenzaldehyde,allylsalicylaldehyde,o-allylsalicylaldehyde
101335
2-prop-2-enoxybenzaldehyde
C=CCOC1=CC=CC=C1C=O
28752-82-1
C10H10O2
162.188
MFCD00014130
BXCJDECTRRMSCV-UHFFFAOYSA-N
2-allyloxy benzaldehyde,2-allyloxybenzaldehyde,o-allyloxy benzaldehyde,o-allyloxybenzaldehyde,benzaldehyde, 2-2-propenyloxy,benzaldehyde, o-allyloxy,2-prop-2-en-1-yloxy benzaldehyde,2-prop-2-enyloxybenzaldehyde,allylsalicylaldehyde,o-allylsalicylaldehyde
101335
2-prop-2-enoxybenzaldehyde
C=CCOC1=CC=CC=C1C=O
Quantity 25mL
Infrared Spectrum Authentic
Refractive Index 1.5540 to 1.5560 (20°C, 589nm)
Packaging Glass bottle
Color Yellow to Orange
Density 1.0790g/mL
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Linear Formula CH2CHCH2OC6H4CHO
Formula Weight 162.19
Physical Form Liquid
Percent Purity ≥88%
Chemical Name or Material 2-Allyloxybenzaldehyde
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Recommended Storage : Product may darken during storage