Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Allyloxybenzaldehyde, 97%

Manufacturer:  Alfa Aesar™ B2511914

Quantity 25g

 View more versions of this product

Catalog No. AAB2511914


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

28752-82-1
162.19
2-Allyloxybenzaldehyde
C10H10O2
MFCD00014130

Specifications

28752-82-1
162.19
129° to 132°C (10mmHg)
>110°C (230°F)
97%
C10H10O2
MFCD00014130
25g
2-Allyloxybenzaldehyde

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit