Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amidinopyridine hydrochloride, 97%, Acros Organics

Click to view available options
Quantity:
5g
50g
51285-26-8
C6H7N3·ClH
157.6
GMHCEDDZKAYPLB-UHFFFAOYSA-N
picolinimidamide hydrochloride,pyridine-2-carboximidamide hydrochloride,2-amidinopyridine hydrochloride,pyridine-2-carboxamidine hydrochloride,picolinamidine hydrochloride,picolinamidine hcl,picolinimidamide hcl,pyridine-2-carboximidamide, hcl,2-amidinopyridine,2-amidinopyridinium chloride
12403615
pyridine-2-carboximidamide;hydrochloride
C1=CC=NC(=C1)C(=N)N.Cl
51285-26-8
C6H7N3·ClH
157.6
GMHCEDDZKAYPLB-UHFFFAOYSA-N
picolinimidamide hydrochloride,pyridine-2-carboximidamide hydrochloride,2-amidinopyridine hydrochloride,pyridine-2-carboxamidine hydrochloride,picolinamidine hydrochloride,picolinamidine hcl,picolinimidamide hcl,pyridine-2-carboximidamide, hcl,2-amidinopyridine,2-amidinopyridinium chloride
12403615
pyridine-2-carboximidamide;hydrochloride
C1=CC=NC(=C1)C(=N)N.Cl
Quantity 50g
Melting Point 146.0°C to 148.0°C
Packaging Glass bottle
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Formula Weight 157.6
Percent Purity 97%
Chemical Name or Material 2-Amidinopyridine hydrochloride