Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-1,1,3-tricyano-1-propene 98.0+%, TCI America™

Click to view available options
Quantity:
25g
250g
868-54-2
C6H4N4
132.126
MFCD00001851
BNHGNFYPZNDLAF-UHFFFAOYSA-N
malononitrile dimer,2-amino-1-propene-1,1,3-tricarbonitrile,2-amino-1,1,3-tricyano-1-propene,2-amino-1,1,3-tricyanopropene,upjohn 9189,tcap,2-amino-1,1,3-propenetricarbonitrile,1,1,3-tricyano-2-aminopropene,1-amino-2-cyanoethylidene malononitrile,tcap van
13356
2-aminoprop-1-ene-1,1,3-tricarbonitrile
C(C#N)C(=C(C#N)C#N)N
868-54-2
C6H4N4
132.126
MFCD00001851
BNHGNFYPZNDLAF-UHFFFAOYSA-N
malononitrile dimer,2-amino-1-propene-1,1,3-tricarbonitrile,2-amino-1,1,3-tricyano-1-propene,2-amino-1,1,3-tricyanopropene,upjohn 9189,tcap,2-amino-1,1,3-propenetricarbonitrile,1,1,3-tricyano-2-aminopropene,1-amino-2-cyanoethylidene malononitrile,tcap van
13356
2-aminoprop-1-ene-1,1,3-tricarbonitrile
C(C#N)C(=C(C#N)C#N)N
Color Undesignated
Melting Point 169°C
Quantity 250g
UN Number 3439
Formula Weight 132.13
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 2-Amino-1,1,3-tricyano-1-propene

RTECSNumber : UD2340000

TSCA : No