Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-1,3-propanediol hydrochloride, 98%

Click to view available options
Quantity:
0.25g
1g
73708-65-3
C3H10ClNO2
127.568
MFCD00012592
VDGBLRKKPTUYNW-UHFFFAOYSA-N
serinol hydrochloride,2-aminopropane-1,3-diol hydrochloride,2-amino-1,3-propanediol hydrochloride,1,3-propanediol, 2-amino-, hydrochloride,dl-serinol,acmc-1bl7f,2-amino-1,3-propanediol hyrochloride
2734815
2-aminopropane-1,3-diol;hydrochloride
C(C(CO)N)O.Cl
73708-65-3
C3H10ClNO2
127.568
MFCD00012592
VDGBLRKKPTUYNW-UHFFFAOYSA-N
serinol hydrochloride,2-aminopropane-1,3-diol hydrochloride,2-amino-1,3-propanediol hydrochloride,1,3-propanediol, 2-amino-, hydrochloride,dl-serinol,acmc-1bl7f,2-amino-1,3-propanediol hyrochloride
2734815
2-aminopropane-1,3-diol;hydrochloride
C(C(CO)N)O.Cl
Quantity 1g
Melting Point 104°C to 106°C
Formula Weight 127.57
Percent Purity 98%
Chemical Name or Material 2-Amino-1,3-propanediol hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning