Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (+/-)-2-Amino-1-butanol, 97%

Click to view available options
Quantity:
100mL
500mL
96-20-8
C4H11NO
89.138
MFCD00008095
JCBPETKZIGVZRE-UHFFFAOYSA-N
2-amino-1-butanol,2-aminobutanol,dl-2-amino-1-butanol,1-butanol, 2-amino,2-aminobutyl alcohol,1-hydroxy-2-butylamine,butanol-2-amine,1-hydroxymethyl propylamine,2-amino-1-hydroxybutane,2-amino-n-butyl alcohol
22129
2-aminobutan-1-ol
CCC(CO)N
96-20-8
C4H11NO
89.138
MFCD00008095
JCBPETKZIGVZRE-UHFFFAOYSA-N
2-amino-1-butanol,2-aminobutanol,dl-2-amino-1-butanol,1-butanol, 2-amino,2-aminobutyl alcohol,1-hydroxy-2-butylamine,butanol-2-amine,1-hydroxymethyl propylamine,2-amino-1-hydroxybutane,2-amino-n-butyl alcohol
22129
2-aminobutan-1-ol
CCC(CO)N
Quantity 100mL
Melting Point -2°C
Boiling Point 176°C to 178°C
pH 11.1
Odor Amine-like
Flash Point 89°C (192°F)
UN Number UN2735
Beilstein 1098274
Merck Index 14,427
Formula Weight 89.14
Percent Purity 97%
Chemical Name or Material (+/-)-2-Amino-1-butanol
Show More Show Less

GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

RTECSNumber : EK9625000

TSCA : TSCA