Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-1-cyclopentene-1-carbonitrile, 98%

Click to view available options
Quantity:
25g
100g
2941-23-3
C6H8N2
108.144
MFCD00517551
NSMYBPIHVACKQG-UHFFFAOYSA-N
2-amino-1-cyclopentene-1-carbonitrile,2-aminocyclopent-1-enecarbonitrile,2-aminocyclopent-1-ene-1-carbonitrile,1-amino-2-cyano-1-cyclopentene,1-cyclopentene-1-carbonitrile, 2-amino,unii-u54887v9pj,2-amino-cyclopent-1-enecarbonitrile,1-amino-2-cyanocyclopent-1-ene,1-amino-2-cyano-1,2-cyclopentene,1-cyclopentene-1-carbonitrile, 2-amino-6ci,7ci,8ci,9ci
287275
2-aminocyclopentene-1-carbonitrile
C1CC(=C(C1)N)C#N
2941-23-3
C6H8N2
108.144
MFCD00517551
NSMYBPIHVACKQG-UHFFFAOYSA-N
2-amino-1-cyclopentene-1-carbonitrile,2-aminocyclopent-1-enecarbonitrile,2-aminocyclopent-1-ene-1-carbonitrile,1-amino-2-cyano-1-cyclopentene,1-cyclopentene-1-carbonitrile, 2-amino,unii-u54887v9pj,2-amino-cyclopent-1-enecarbonitrile,1-amino-2-cyanocyclopent-1-ene,1-amino-2-cyano-1,2-cyclopentene,1-cyclopentene-1-carbonitrile, 2-amino-6ci,7ci,8ci,9ci
287275
2-aminocyclopentene-1-carbonitrile
C1CC(=C(C1)N)C#N
Applications
2-Amino-1-cyclopentene-1-carbonitrile s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Incompatible with heat and oxidizing agents.
Quantity 100g
Melting Point 147°C to 148°C
Color White
UN Number UN3439
Beilstein 742061
Formula Weight 108.14
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Amino-1-cyclopentene-1-carbonitrile

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

RTECSNumber : GY5976010

storageNote1 : 2° to 8°C