Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-1-cyclopentene-1-carbonitrile, 98%, Thermo Scientific Chemicals

Catalog Number AAH6621914
Click to view available options
Quantity:
25 g
100 g
2941-23-3
C6H8N2
108.144
MFCD00517551
NSMYBPIHVACKQG-UHFFFAOYSA-N
2-amino-1-cyclopentene-1-carbonitrile, 2-aminocyclopent-1-enecarbonitrile, 2-aminocyclopent-1-ene-1-carbonitrile, 1-amino-2-cyano-1-cyclopentene, 1-cyclopentene-1-carbonitrile, 2-amino, unii-u54887v9pj, 2-amino-cyclopent-1-enecarbonitrile, 1-amino-2-cyanocyclopent-1-ene, 1-amino-2-cyano-1,2-cyclopentene, 1-cyclopentene-1-carbonitrile, 2-amino-6ci,7ci,8ci,9ci
287275
2-aminocyclopentene-1-carbonitrile
C1CC(=C(C1)N)C#N
2941-23-3
C6H8N2
108.144
MFCD00517551
NSMYBPIHVACKQG-UHFFFAOYSA-N
2-amino-1-cyclopentene-1-carbonitrile, 2-aminocyclopent-1-enecarbonitrile, 2-aminocyclopent-1-ene-1-carbonitrile, 1-amino-2-cyano-1-cyclopentene, 1-cyclopentene-1-carbonitrile, 2-amino, unii-u54887v9pj, 2-amino-cyclopent-1-enecarbonitrile, 1-amino-2-cyanocyclopent-1-ene, 1-amino-2-cyano-1,2-cyclopentene, 1-cyclopentene-1-carbonitrile, 2-amino-6ci,7ci,8ci,9ci
287275
2-aminocyclopentene-1-carbonitrile
C1CC(=C(C1)N)C#N

2-Amino-1-cyclopentene-1-carbonitrile s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-1-cyclopentene-1-carbonitrile s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 147°C to 148°C
Color White
Quantity 25 g
UN Number UN3439
Beilstein 742061
Solubility Information Slightly soluble in water.
Formula Weight 108.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Amino-1-cyclopentene-1-carbonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
RTECSNumber GY5976010
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit