Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-2′,4-dichlorodiphenyl Ether 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
56966-48-4
C12H9Cl2NO
254.11
MFCD00025218
PHORTNLNXNZNET-UHFFFAOYSA-N
5-Chloro-2-(2-chlorophenoxy)aniline
92601
5-chloro-2-(2-chlorophenoxy)aniline
C1=CC=C(C(=C1)OC2=C(C=C(C=C2)Cl)N)Cl
56966-48-4
C12H9Cl2NO
254.11
MFCD00025218
PHORTNLNXNZNET-UHFFFAOYSA-N
5-Chloro-2-(2-chlorophenoxy)aniline
92601
5-chloro-2-(2-chlorophenoxy)aniline
C1=CC=C(C(=C1)OC2=C(C=C(C=C2)Cl)N)Cl
Boiling Point 210°C
Color Undesignated
Quantity 25g
Formula Weight 254.11
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2-Amino-2′,4-dichlorodiphenyl Ether