Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-2',5-dichlorobenzophenone 99%, Acros Organics

Click to view available options
Quantity:
50g
Packaging:
Glass bottle
2958-36-3
C13H9Cl2NO
266.121
MFCD00007840
KWZYIAJRFJVQDO-UHFFFAOYSA-N
2-amino-2',5-dichlorobenzophenone,2-amino-5-chlorophenyl 2-chlorophenyl methanone,4-chloro-2-2-chlorobenzoyl aniline,methanone, 2-amino-5-chlorophenyl 2-chlorophenyl,unii-g806veo3ke,2-amino-5-chloro-2'-chlorobenzophenone,2-amino-5-chlorophenyl-2-chlorophenyl methanone,g806veo3ke,benzophenone, 2-amino-2',5-dichloro,2-amino-5,2'-dichlorobenzophenone
18069
(2-amino-5-chlorophenyl)-(2-chlorophenyl)methanone
C1=CC=C(C(=C1)C(=O)C2=C(C=CC(=C2)Cl)N)Cl
2958-36-3
C13H9Cl2NO
266.121
MFCD00007840
KWZYIAJRFJVQDO-UHFFFAOYSA-N
2-amino-2',5-dichlorobenzophenone,2-amino-5-chlorophenyl 2-chlorophenyl methanone,4-chloro-2-2-chlorobenzoyl aniline,methanone, 2-amino-5-chlorophenyl 2-chlorophenyl,unii-g806veo3ke,2-amino-5-chloro-2'-chlorobenzophenone,2-amino-5-chlorophenyl-2-chlorophenyl methanone,g806veo3ke,benzophenone, 2-amino-2',5-dichloro,2-amino-5,2'-dichlorobenzophenone
18069
(2-amino-5-chlorophenyl)-(2-chlorophenyl)methanone
C1=CC=C(C(=C1)C(=O)C2=C(C=CC(=C2)Cl)N)Cl
Quantity 50g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Undesignated
Melting Point 86°C to 89°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula H2NC6H3(Cl)COC6H4Cl
Formula Weight 266.13
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material 2-Amino-2', 5-dichlorobenzophenone, 99%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.