Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-2'-chloro-5-nitrobenzophenone, 98%

Click to view available options
Quantity:
5g
25g
100g
2011-66-7
C13H9ClN2O3
276.676
MFCD00792453
GRDGBWVSVMLKBV-UHFFFAOYSA-N
2-amino-2'-chloro-5-nitrobenzophenone,2-amino-5-nitrophenyl 2-chlorophenyl methanone,2-amino-5-nitro-2'-chlorobenzophenone,2-amino-2'-chloro-5-nitro benzophenone,2-2-chlorobenzoyl-4-nitroaniline,methanone, 2-amino-5-nitrophenyl 2-chlorophenyl,2-amino-5-nitrophenyl-2-chlorophenyl methanone,2-amino-5-nitro-phenyl-2-chloro-phenyl-methanone,2-amino-5-nitro-2'-chloro benzophenone,pubchem3255
74830
(2-amino-5-nitrophenyl)-(2-chlorophenyl)methanone
C1=CC=C(C(=C1)C(=O)C2=C(C=CC(=C2)[N+](=O)[O-])N)Cl
2011-66-7
C13H9ClN2O3
276.676
MFCD00792453
GRDGBWVSVMLKBV-UHFFFAOYSA-N
2-amino-2'-chloro-5-nitrobenzophenone,2-amino-5-nitrophenyl 2-chlorophenyl methanone,2-amino-5-nitro-2'-chlorobenzophenone,2-amino-2'-chloro-5-nitro benzophenone,2-2-chlorobenzoyl-4-nitroaniline,methanone, 2-amino-5-nitrophenyl 2-chlorophenyl,2-amino-5-nitrophenyl-2-chlorophenyl methanone,2-amino-5-nitro-phenyl-2-chloro-phenyl-methanone,2-amino-5-nitro-2'-chloro benzophenone,pubchem3255
74830
(2-amino-5-nitrophenyl)-(2-chlorophenyl)methanone
C1=CC=C(C(=C1)C(=O)C2=C(C=CC(=C2)[N+](=O)[O-])N)Cl
Quantity 100g
Sensitivity Air Sensitive
Color Yellow
Melting Point 119°C to 123°C
Beilstein 2814216
Formula Weight 276.68
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2-Amino-2'-chloro-5-nitrobenzophenone

storageNote1 : 2° to 8°C