Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-2-cyanoacetamide, 97+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
6719-21-7
C3H5N3O
99.09
MFCD00040532
JRWAUKYINYWSTA-UHFFFAOYSA-N
aminocyanoacetamide,3-nitriloalaninamide,acetamide, 2-amino-2-cyano,2-aminocyanoacetamide,aminocyanacetamide,2-amino-cyanoacetamide,2-amino-2cyanoacetamide,a-amino-a-cyanoacetamide,2-amino-2-cyanacetamide,2-amino 2-cyano acetamide
98341
2-amino-2-cyanoacetamide
C(#N)C(C(=O)N)N
6719-21-7
C3H5N3O
99.09
MFCD00040532
JRWAUKYINYWSTA-UHFFFAOYSA-N
aminocyanoacetamide,3-nitriloalaninamide,acetamide, 2-amino-2-cyano,2-aminocyanoacetamide,aminocyanacetamide,2-amino-cyanoacetamide,2-amino-2cyanoacetamide,a-amino-a-cyanoacetamide,2-amino-2-cyanacetamide,2-amino 2-cyano acetamide
98341
2-amino-2-cyanoacetamide
C(#N)C(C(=O)N)N
Melting Point 120°C to 124°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color Beige to Tan
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Linear Formula NCCH(NH2)CONH2
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ether and methanol
Formula Weight 99.09
Physical Form Powder
Percent Purity 97+%
Chemical Name or Material 2-Amino-2-cyanoacetamide
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 229-771-3