Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2-Amino-2-ethyl-1,3-propanediol, 97%
Click to view available options
Quantity:
100g
500g
Description
Applications
It is used in pharmaceutical industry and in medicine.
Solubility
Fully miscible in water.
Notes
Moisture sensitive. Store at room temperature. Incompatible with oxidizing agents.
It is used in pharmaceutical industry and in medicine.
Solubility
Fully miscible in water.
Notes
Moisture sensitive. Store at room temperature. Incompatible with oxidizing agents.
Specifications
Refractive Index | 1.488 |
Density | 1.099 |
Sensitivity | Hygroscopic |
Boiling Point | 152°C to 153°C (10mmHg) |
Quantity | 100g |
Flash Point | >110°C (230°F) |
Merck Index | 14,439 |
Solubility Information | Fully miscible in water. |
Formula Weight | 119.16 |
Percent Purity | 97% |
Show More |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P280-P305+P351+P338-P310
H318
EINECSNumber : 204-101-2
TSCA : Yes
Recommended Storage : Ambient temperatures