Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3,5-dibromobenzotrifluoride, 98%, Acros Organics

Click to view available options
Quantity:
5g
71757-14-7
C7H4Br2F3N
318.919
ZYBKXMTWWFTYKN-UHFFFAOYSA-N
2,4-dibromo-6-trifluoromethyl aniline,2-amino-3,5-dibromobenzotrifluoride,benzenamine,2,4-dibromo-6-trifluoromethyl,2,4-dibromo-6-trifluoromethyl-phenylamine,4,6-dibromo-2-trifluoromethyl phenylamine,pubchem2759,acmc-209ojz,buttpark 29\06-98,zerenex e/9071924,2,4-bis bromanyl-6-trifluoromethyl aniline
737213
2,4-dibromo-6-(trifluoromethyl)aniline
C1=C(C=C(C(=C1Br)N)C(F)(F)F)Br
71757-14-7
C7H4Br2F3N
318.919
ZYBKXMTWWFTYKN-UHFFFAOYSA-N
2,4-dibromo-6-trifluoromethyl aniline,2-amino-3,5-dibromobenzotrifluoride,benzenamine,2,4-dibromo-6-trifluoromethyl,2,4-dibromo-6-trifluoromethyl-phenylamine,4,6-dibromo-2-trifluoromethyl phenylamine,pubchem2759,acmc-209ojz,buttpark 29\06-98,zerenex e/9071924,2,4-bis bromanyl-6-trifluoromethyl aniline
737213
2,4-dibromo-6-(trifluoromethyl)aniline
C1=C(C=C(C(=C1Br)N)C(F)(F)F)Br
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 318.92
Chemical Name or Material 2-Amino-3, 5-dibromobenzotrifluoride, 98%

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning