Alfa Aesar™ 2-Amino-3,5-dibromopyrazine, 99%

Manufacturer: Alfa Aesar™ H28744MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH28744MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24241-18-7
    Molecular Formula C4H3Br2N3
    Formula Weight 252.9
    MDL Number MFCD00673150
    Synonym 3,5-Dibromo-2-pyrazinamine
    Chemical Name or Material 2-Amino-3,5-dibromopyrazine

    Specifications

    Formula Weight 252.9
    Quantity 250mg
    MDL Number MFCD00673150
    Melting Point 114° to 117°C
    Molecular Formula C4H3Br2N3
    CAS 24241-18-7
    Synonym 3,5-Dibromo-2-pyrazinamine
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 2-Amino-3,5-dibromopyrazine
    Assay 99%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit