Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-3,5-dichlorobenzoic acid, 97%

Click to view available options
Quantity:
10g
50g
250g
2789-92-6
C7H5Cl2NO2
206.022
MFCD00017088
KTHTXLUIEAIGCD-UHFFFAOYSA-N
3,5-dichloroanthranilic acid,benzoic acid, 2-amino-3,5-dichloro,anthranilic acid, 3,5-dichloro,3,5-dichloro-2-aminobenzoic acid,2-amino-3,5-dichloro-benzoic acid,timtec-bb sbb003493,2-amino-3,5-dichlorobenzoicacid,acmc-209gzl,rarechem al bo 0770,buttpark 49\07-46
76036
2-amino-3,5-dichlorobenzoic acid
C1=C(C=C(C(=C1Cl)N)C(=O)O)Cl
2789-92-6
C7H5Cl2NO2
206.022
MFCD00017088
KTHTXLUIEAIGCD-UHFFFAOYSA-N
3,5-dichloroanthranilic acid,benzoic acid, 2-amino-3,5-dichloro,anthranilic acid, 3,5-dichloro,3,5-dichloro-2-aminobenzoic acid,2-amino-3,5-dichloro-benzoic acid,timtec-bb sbb003493,2-amino-3,5-dichlorobenzoicacid,acmc-209gzl,rarechem al bo 0770,buttpark 49\07-46
76036
2-amino-3,5-dichlorobenzoic acid
C1=C(C=C(C(=C1Cl)N)C(=O)O)Cl
Applications
2-Amino-3,5-dichlorobenzoic acid, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
This material is stable if stored under proper conditions. Avoid excessive heat and light. Keep away from strong oxidizing agents.
Quantity 10g
Melting Point 225°C to 227°C
Beilstein 779100
Solubility Information Insoluble in water.
Formula Weight 206.03
Percent Purity 97%
Chemical Name or Material 2-Amino-3,5-dichlorobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 220-520-3

RTECSNumber : CB2800000

TSCA : No

Recommended Storage : Ambient temperatures