Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-3,5-dichlorobenzonitrile, 97%

Click to view available options
Quantity:
1g
5g
36764-94-0
C7H4Cl2N2
187.023
MFCD00158947
ZHKNDJRPOVUPMT-UHFFFAOYSA-N
2-amino-3,5-dichloro-benzonitrile,2-amino-3,5-dichlorobenzenecarbonitrile,pubchem11850,3,5-dichloroanthranilonitrile,buttpark 52\14-89,benzonitrile,2-amino-3,5-dichloro,2-amino-3,5-dichlorobenzonitrile
737215
2-amino-3,5-dichlorobenzonitrile
C1=C(C=C(C(=C1Cl)N)C#N)Cl
36764-94-0
C7H4Cl2N2
187.023
MFCD00158947
ZHKNDJRPOVUPMT-UHFFFAOYSA-N
2-amino-3,5-dichloro-benzonitrile,2-amino-3,5-dichlorobenzenecarbonitrile,pubchem11850,3,5-dichloroanthranilonitrile,buttpark 52\14-89,benzonitrile,2-amino-3,5-dichloro,2-amino-3,5-dichlorobenzonitrile
737215
2-amino-3,5-dichlorobenzonitrile
C1=C(C=C(C(=C1Cl)N)C#N)Cl
Melting Point 123°C to 127°C (literature)
Quantity 1g
Beilstein 2720575
Formula Weight 187.02
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Amino-3,5-dichlorobenzonitrile

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning