Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3,5-dichlorobenzotrifluoride, 99%, Acros Organics

Click to view available options
Quantity:
1g
62593-17-3
C7H4Cl2F3N
230.011
MFCD00174313
OCJPQZFOBGQYEA-UHFFFAOYSA-N
2,4-dichloro-6-trifluoromethyl aniline,2-amino-3,5-dichlorobenzotrifluoride,benzenamine, 2,4-dichloro-6-trifluoromethyl,2,4-dichloro-6-trifluoromethyl phenylamine,3,5-dichloro-2-aminobenzotrifluoride,pubchem2760,2,4-dichloro-6-trifluoromethylanilin,2,4-dichloro-6-trifluoromethyl anilin,2,4-dichloro-6-trifluoromethyl benzenamine
1268255
2,4-dichloro-6-(trifluoromethyl)aniline
C1=C(C=C(C(=C1Cl)N)C(F)(F)F)Cl
62593-17-3
C7H4Cl2F3N
230.011
MFCD00174313
OCJPQZFOBGQYEA-UHFFFAOYSA-N
2,4-dichloro-6-trifluoromethyl aniline,2-amino-3,5-dichlorobenzotrifluoride,benzenamine, 2,4-dichloro-6-trifluoromethyl,2,4-dichloro-6-trifluoromethyl phenylamine,3,5-dichloro-2-aminobenzotrifluoride,pubchem2760,2,4-dichloro-6-trifluoromethylanilin,2,4-dichloro-6-trifluoromethyl anilin,2,4-dichloro-6-trifluoromethyl benzenamine
1268255
2,4-dichloro-6-(trifluoromethyl)aniline
C1=C(C=C(C(=C1Cl)N)C(F)(F)F)Cl
Refractive Index 1.5200 to 1.5220 (20°C, 589nm)
Color Orange
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 230.07
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2-Amino-3, 5-dichlorobenzotrifluoride, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning