Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-3,5-dichloropyridine, 98%

Click to view available options
Quantity:
5g
25g
100g
4214-74-8
C5H4Cl2N2
163.001
MFCD00006313
OCWBGKZFOYMCCN-UHFFFAOYSA-N
2-amino-3,5-dichloropyridine,3,5-dichloro-2-pyridinamine,2-pyridinamine, 3,5-dichloro,3,5-dichloro-3-pyridylamine,3,5-dichloro-2-aminopyridine,3,5-dichloro-2-pyridylamine,3,5-dichloropyridin-2-ylamine,pyridine, 2-amino-3,5-dichloro,pubchem13504,acmc-209jnv
77886
3,5-dichloropyridin-2-amine
C1=C(C=NC(=C1Cl)N)Cl
4214-74-8
C5H4Cl2N2
163.001
MFCD00006313
OCWBGKZFOYMCCN-UHFFFAOYSA-N
2-amino-3,5-dichloropyridine,3,5-dichloro-2-pyridinamine,2-pyridinamine, 3,5-dichloro,3,5-dichloro-3-pyridylamine,3,5-dichloro-2-aminopyridine,3,5-dichloro-2-pyridylamine,3,5-dichloropyridin-2-ylamine,pyridine, 2-amino-3,5-dichloro,pubchem13504,acmc-209jnv
77886
3,5-dichloropyridin-2-amine
C1=C(C=NC(=C1Cl)N)Cl
Quantity 5g
Odor Unpleasant
Melting Point 81°C to 83°C
Beilstein 119376
Formula Weight 163.01
Percent Purity 98%
Chemical Name or Material 2-Amino-3,5-dichloropyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning