Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-benzyloxypyridine, 98.5%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
24016-03-3
C12H12N2O
200.24
MFCD00006316
NMCBWICNRJLKKM-UHFFFAOYSA-N
2-amino-3-benzyloxypyridine,3-benzyloxy pyridin-2-amine,3-benzyloxy-2-pyridylamine,2-pyridinamine, 3-phenylmethoxy,2-amino-3-benzyloxy pyridine,3ip,3-phenylmethoxy-2-pyridylamine,3-benzyloxy pyridin-2-amin,3fty
90334
3-phenylmethoxypyridin-2-amine
C1=CC=C(C=C1)COC2=C(N=CC=C2)N
24016-03-3
C12H12N2O
200.24
MFCD00006316
NMCBWICNRJLKKM-UHFFFAOYSA-N
2-amino-3-benzyloxypyridine,3-benzyloxy pyridin-2-amine,3-benzyloxy-2-pyridylamine,2-pyridinamine, 3-phenylmethoxy,2-amino-3-benzyloxy pyridine,3ip,3-phenylmethoxy-2-pyridylamine,3-benzyloxy pyridin-2-amin,3fty
90334
3-phenylmethoxypyridin-2-amine
C1=CC=C(C=C1)COC2=C(N=CC=C2)N
Quantity 25g
Infrared Spectrum Authentic
Color Brown to Green
Packaging Glass bottle
Melting Point 94°C to 98°C
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Formula Weight 200.24
Physical Form Crystalline Powder
Percent Purity 98.50%
Chemical Name or Material 2-Amino-3-benzyloxypyridine, 98.5%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 245-983-9

TSCA : TSCA